ENA428682784 Enamine

2-(4-chlorophenyl)-5-ethyl-1H-imidazole-4-carboxylic acid

  • MDL number MFCD12411561
  • Cas Number : 1156729-24-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-180846

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180846
Availability details for
Quantity
  • Availability
  • Add to Cart