ENA428682789 Enamine

[(2,3-dihydro-1-benzofuran-5-yl)methyl](methyl)amine hydrochloride

  • MDL number MFCD28118315
  • Cas Number : 1645513-87-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-180914

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180914
Availability details for
Quantity
  • Availability
  • Add to Cart