ENA428682794 Enamine

(3-tert-butyl-1,2,4-oxadiazol-5-yl)methanol

  • MDL number MFCD12138059
  • Cas Number : 1153757-34-6
  • Molecular Weight : 156.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-180937

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-180937
Availability details for
Quantity
  • Availability
  • Add to Cart