ENA428682810 Enamine

4-formyl-2-methylbenzonitrile

  • MDL number MFCD18394298
  • Cas Number : 27613-35-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-181018

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181018
Availability details for
Quantity
  • Availability
  • Add to Cart