ENA428682814 Enamine

1-(1-methyl-1H-pyrazol-5-yl)piperidin-3-amine

  • MDL number MFCD19620828
  • Cas Number : 1342851-15-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-181103
Availability details for
Quantity
  • Availability
  • Add to Cart