ENA428682821 Enamine

2-(diethylamino)-6H,7H-[1,3]thiazolo[4,5-d]pyrimidin-7-one

  • MDL number MFCD09041400
  • Cas Number : 926194-82-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-181194
Availability details for
Quantity
  • Availability
  • Add to Cart