ENA428682822 Enamine

2-(pyrrolidin-1-yl)-6H,7H-[1,3]thiazolo[4,5-d]pyrimidin-7-one

  • MDL number MFCD09041752
  • Cas Number : 99967-86-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-181195

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181195
Availability details for
Quantity
  • Availability
  • Add to Cart