ENA428682825 Enamine

5-(4-bromophenyl)-2,3-dihydro-1,3-oxazole-2-thione

  • MDL number MFCD14703283
  • Cas Number : 931966-96-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-181252

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181252
Availability details for
Quantity
  • Availability
  • Add to Cart