ENA428682826 Enamine

2-(piperidin-1-yl)-6H,7H-[1,3]thiazolo[4,5-d]pyrimidin-7-one

  • MDL number MFCD14703290
  • Cas Number : 99967-87-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 305-307
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-181254

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181254
Availability details for
Quantity
  • Availability
  • Add to Cart