ENA428682828 Enamine

6-[4-(2-ethoxyphenyl)piperazin-1-yl]-2,3-dihydropyridazine-3-thione

  • MDL number MFCD14729822
  • Molecular Weight : 316.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-181268

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181268
Availability details for
Quantity
  • Availability
  • Add to Cart