ENA428682829 Enamine

6-[4-(3-methylphenyl)piperazin-1-yl]-2,3-dihydropyridazine-3-thione

  • MDL number MFCD20231831
  • Cas Number : 100241-61-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-181281

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181281
Availability details for
Quantity
  • Availability
  • Add to Cart