ENA428682847 Enamine

methyl 6-azaspiro[3.4]octane-8-carboxylate

  • MDL number MFCD28118197
  • Cas Number : 1597285-39-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-151909

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-151909
Availability details for
Quantity
  • Availability
  • Add to Cart