ENA428682872 Enamine

2-amino-4-chloro-3-fluorobenzoic acid

  • MDL number MFCD11193652
  • Cas Number : 1039878-71-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-181503

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181503
Availability details for
Quantity
  • Availability
  • Add to Cart