ENA428682876 Enamine

1-(2,6-dichlorophenyl)cyclobutane-1-carboxylic acid

  • MDL number MFCD11037041
  • Cas Number : 1039951-74-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-181655

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181655
Availability details for
Quantity
  • Availability
  • Add to Cart