ENA428682883 Enamine

(2-chloro-4-methylphenyl)methanol

  • MDL number MFCD18393674
  • Cas Number : 39652-31-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-181803

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181803
Availability details for
Quantity
  • Availability
  • Add to Cart