ENA428682893 Enamine

4,5,6,7-tetrahydro-1H-1,3-benzodiazole-4-carboxylic acid

  • MDL number MFCD18807882
  • Cas Number : 361395-33-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-181939

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181939
Availability details for
Quantity
  • Availability
  • Add to Cart