ENA428682922 Enamine

5-bromo-3-[(1,2,4-oxadiazol-3-yl)methyl]-3,4-dihydropyrimidin-4-one

  • MDL number MFCD21746747
  • Cas Number : 1540818-93-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-182366

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182366
Availability details for
Quantity
  • Availability
  • Add to Cart