ENA428682926 Enamine

N-(2-aminoethyl)-N,3-dimethylaniline

  • MDL number MFCD10031337
  • Cas Number : 101374-10-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-182401

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182401
Availability details for
Quantity
  • Availability
  • Add to Cart