ENA428682931 Enamine

3-(1-methyl-1H-imidazol-2-yl)piperidine

  • MDL number MFCD21909872
  • Cas Number : 933701-83-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-182438

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182438
Availability details for
Quantity
  • Availability
  • Add to Cart