ENA428682936 Enamine

3-(5-methylfuran-2-yl)-3-oxopropanenitrile

  • MDL number MFCD11170514
  • Cas Number : 197250-40-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-182467
Availability details for
Quantity
  • Availability
  • Add to Cart