ENA428682946 Enamine

(but-3-yn-2-yl)(methyl)amine

  • MDL number MFCD00041666
  • Cas Number : 4070-90-0
  • Molecular Weight : 83.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-182523

Safety Information

signalword Danger
pcodes P210,P233,P240,P241,P242,P243,P260,P264,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P363,P370+P378,P403+P235,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-182523
pictograms GHS02,GHS05
hstatements H225,H314
Availability details for
Quantity
  • Availability
  • Add to Cart