ENA428682953 Enamine

N2-phenyl-1,3,4-oxadiazole-2,5-diamine

  • MDL number MFCD20693988
  • Cas Number : 74099-09-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-182562

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182562
Availability details for
Quantity
  • Availability
  • Add to Cart