ENA428682954 Enamine

2-(1-methylcyclopropyl)pyrimidin-4-amine

  • MDL number MFCD23906901
  • Cas Number : 1521482-21-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-182564

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182564
Availability details for
Quantity
  • Availability
  • Add to Cart