ENA428682959 Enamine

5-iodothiophene-3-carboxamide

  • MDL number MFCD23722930
  • Cas Number : 18800-08-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-182587

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182587
Availability details for
Quantity
  • Availability
  • Add to Cart