ENA428682962 Enamine

2-chloro-N-hydroxybenzene-1-carbonimidoyl chloride

  • MDL number MFCD22377764
  • Cas Number : 1383929-39-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 57-58
Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-182609

Safety Information

signalword Danger
pcodes P264,P270,P301+P310,P330,P405,P501
hcodes T
MSDS https://www.enaminestore.com/catalog/EN300-182609
pictograms GHS06
hstatements H301
Availability details for
Quantity
  • Availability
  • Add to Cart