ENA428682984 Enamine

1-(1H-imidazol-2-yl)piperazine dihydrochloride

  • MDL number MFCD08669026
  • Cas Number : 1795503-50-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-182661

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182661
Availability details for
Quantity
  • Availability
  • Add to Cart