ENA428682994 Enamine

2-(4-fluorophenyl)-1-phenylethan-1-one

  • MDL number MFCD02260684
  • Cas Number : 347-91-1
  • Molecular Weight : 214.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-182711

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182711
Availability details for
Quantity
  • Availability
  • Add to Cart