ENA428682995 Enamine

methyl 2-[(2-methyl-1,3-thiazol-4-yl)methyl]-1H-imidazole-4-carboxylate

  • MDL number MFCD17224849
  • Cas Number : 1341188-09-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-182712

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182712
Availability details for
Quantity
  • Availability
  • Add to Cart