ENA428682996 Enamine

3-bromo-4-(4-chloro-2-fluorophenyl)butan-2-one

  • MDL number MFCD16748562
  • Cas Number : 1250030-38-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-182714

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182714
Availability details for
Quantity
  • Availability
  • Add to Cart