ENA428682998 Enamine

1-[(3-fluorophenyl)methyl]-5-methyl-1H-1,2,3-triazol-4-amine

  • MDL number MFCD21947924
  • Cas Number : 1507906-31-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-182718

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182718
Availability details for
Quantity
  • Availability
  • Add to Cart