ENA428683001 Enamine

2-(3,3-dimethylpiperidin-1-yl)ethan-1-amine

  • MDL number MFCD19643333
  • Cas Number : 34970-93-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-182729

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182729
Availability details for
Quantity
  • Availability
  • Add to Cart