ENA428683004 Enamine

3-[5-(trifluoromethyl)-1H-imidazol-2-yl]piperidine

  • MDL number MFCD12105851
  • Cas Number : 1153265-18-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-182748

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182748
Availability details for
Quantity
  • Availability
  • Add to Cart