ENA428683007 Enamine

2-hydroxy-1,3-thiazole-5-carbaldehyde

  • MDL number MFCD24110457
  • Cas Number : 1489487-02-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-182753
Availability details for
Quantity
  • Availability
  • Add to Cart