ENA428683014 Enamine

methyl 3-[(2-methoxy-2-oxoethyl)sulfanyl]-2-methylpropanoate

  • MDL number MFCD12144490
  • Cas Number : 38293-65-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-182837

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182837
Availability details for
Quantity
  • Availability
  • Add to Cart