ENA428683017 Enamine

4-bromo-1-{1,9-dioxaspiro[5.5]undecan-4-yl}-1H-pyrazole

  • MDL number MFCD24780411
  • Cas Number : 1499223-80-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-182843

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182843
Availability details for
Quantity
  • Availability
  • Add to Cart