ENA428683026 Enamine

1-(2-isocyanatoethyl)-3,5-dimethoxybenzene

  • MDL number MFCD03701577
  • Cas Number : 480439-01-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-182886

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182886
s_phrases_eu 23-26-36
Availability details for
Quantity
  • Availability
  • Add to Cart