ENA428683032 Enamine

2-methyl-4-(trifluoromethyl)phenol

  • MDL number MFCD16997488
  • Cas Number : 77227-90-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-182897

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182897
Availability details for
Quantity
  • Availability
  • Add to Cart