ENA428683033 Enamine

4-(ethanesulfonyl)piperidine hydrochloride

  • MDL number MFCD16294581
  • Cas Number : 1147107-63-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-182898

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182898
Availability details for
Quantity
  • Availability
  • Add to Cart