ENA428683071 Enamine

4-chloro-2-methoxybenzonitrile

  • MDL number MFCD09865166
  • Cas Number : 100960-68-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 95-97
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-182993

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-182993
Availability details for
Quantity
  • Availability
  • Add to Cart