ENA428683075 Enamine

2,2-difluoro-1-(oxolan-2-yl)ethan-1-ol

  • MDL number MFCD23943635
  • Cas Number : 1548587-39-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-183029

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-183029
Availability details for
Quantity
  • Availability
  • Add to Cart