ENA428683077 Enamine

ethyl 4-(4-cyanophenyl)-2,4-dioxobutanoate

  • MDL number MFCD03411527
  • Cas Number : 649557-55-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-183031

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-183031
Availability details for
Quantity
  • Availability
  • Add to Cart