ENA428683081 Enamine

2-methyl-2,8-diazaspiro[4.5]decan-1-one hydrochloride

  • MDL number MFCD02179152
  • Cas Number : 848580-34-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-183042

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-183042
Availability details for
Quantity
  • Availability
  • Add to Cart