ENA428683083 Enamine

3-(4-methyl-1,3-thiazol-2-yl)propanoic acid

  • MDL number MFCD11043594
  • Cas Number : 89776-65-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-183047

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-183047
Availability details for
Quantity
  • Availability
  • Add to Cart