ENA428683085 Enamine

ethyl 3-(4-chlorophenyl)-2-cyanobut-2-enoate

  • Cas Number : 13455-81-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-183076

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-183076
Availability details for
Quantity
  • Availability
  • Add to Cart