ENA428683089 Enamine

methyl 5-methyl-4-nitro-1H-pyrazole-3-carboxylate

  • MDL number MFCD08693983
  • Cas Number : 27116-86-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 134-137
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-183114

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-183114
Availability details for
Quantity
  • Availability
  • Add to Cart