ENA428683092 Enamine

2-bromo-5-(1,3-dioxolan-2-yl)-1,3-thiazole

  • MDL number MFCD06210128
  • Cas Number : 773088-29-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-183118

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-183118
Availability details for
Quantity
  • Availability
  • Add to Cart