ENA428683093 Enamine

3-(propan-2-yl)-2,3-dihydro-1H-inden-1-one

  • MDL number MFCD16301548
  • Cas Number : 22531-07-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-183120

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-183120
Availability details for
Quantity
  • Availability
  • Add to Cart