ENA428683095 Enamine

3-bromo-4-(3-chloro-2-fluorophenyl)butan-2-one

  • MDL number MFCD16748640
  • Cas Number : 1251242-72-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-183125

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-183125
Availability details for
Quantity
  • Availability
  • Add to Cart