ENA428683097 Enamine

4-(pyridin-3-yl)butanoic acid hydrochloride

  • MDL number MFCD18802935
  • Cas Number : 176446-91-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 115-120
Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-183128

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-183128
Availability details for
Quantity
  • Availability
  • Add to Cart