ENA428683098 Enamine

4-chloro-5-iodo-6-(2-methylpropyl)pyrimidine

  • MDL number MFCD23898486
  • Cas Number : 1514650-08-8
  • Molecular Weight : 296.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-183129

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-183129
Availability details for
Quantity
  • Availability
  • Add to Cart